Merck
  • Synthesis, Bottom up Assembly and Thermoelectric Properties of Sb-Doped PbS Nanocrystal Building Blocks.
Synthesis, Bottom up Assembly and Thermoelectric Properties of Sb-Doped PbS Nanocrystal Building Blocks.
Materials (Basel, Switzerland) (2021-02-14)
Doris Cadavid, Kaya Wei, Yu Liu, Yu Zhang, Mengyao Li, Aziz Genç, Taisiia Berestok, Maria Ibáñez, Alexey Shavel, George S Nolas, Andreu Cabot
ABSTRACT
The precise engineering of thermoelectric materials using nanocrystals as their building blocks has proven to be an excellent strategy to increase energy conversion efficiency. Here we present a synthetic route to produce Sb-doped PbS colloidal nanoparticles. These nanoparticles are then consolidated into nanocrystalline PbS:Sb using spark plasma sintering. We demonstrate that the introduction of Sb significantly influences the size, geometry, crystal lattice and especially the carrier concentration of PbS. The increase of charge carrier concentration achieved with the introduction of Sb translates into an increase of the electrical and thermal conductivities and a decrease of the Seebeck coefficient. Overall, PbS:Sb nanomaterial were characterized by two-fold higher thermoelectric figures of merit than undoped PbS.
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Sodium sulfide nonahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
1-Octadecene, technical grade, 90%