Merck
  • Evaluation of human neutrophil elastase inhibitory effect of iridoid glycosides from Hedyotis diffusa.
Evaluation of human neutrophil elastase inhibitory effect of iridoid glycosides from Hedyotis diffusa.
Bioorganic & medicinal chemistry letters (2009-12-17)
Guang-Hua Xu, Young-Hee Kim, Seung-Wook Chi, Soo-Jin Choo, In-Ja Ryoo, Jong-Seog Ahn, Ick-Dong Yoo
ABSTRACT
Five iridoid glycosides were isolated from the MeOH extract of Hedyotis diffusa, and their structures were elucidated as E-6-O-p-methoxycinnamoyl scandoside methyl ester (1), Z-6-O-p-methoxycinnamoyl scandoside methyl ester (2), E-6-O-p-feruloyl scandoside methyl ester (3), E-6-O-p-coumaroyl scandoside methyl ester (4), and Z-6-O-p-coumaroyl scandoside methyl ester (5) by interpretation of their spectroscopic data. All the isolated compounds were evaluated for human neutrophil elastase inhibitory effect, and compound 1 showed potent activity with an IC(50) value of 18.0muM. The molecular docking simulation suggested a structural model for the inhibition of human neutrophil elastase by compound 1.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%