Merck
  • Impact of sequential enzymatic hydrolysis on antioxidant activity and peptide profile of casein hydrolysate.
Impact of sequential enzymatic hydrolysis on antioxidant activity and peptide profile of casein hydrolysate.
Journal of food science and technology (2020-10-23)
Priyanka Singh Rao, Rajesh Bajaj, Bimlesh Mann
ABSTRACT
This paper shows the potential of dual enzyme approach on antioxidant activity of casein hydrolysates. Casein was hydrolysed using the proteolytic enzymes alcalase, flavourzyme in isolation and in sequential order. Casein hydrolysates were evaluated for the degree of hydrolysis, antioxidant activity, molecular weight distribution patterns and peptide sequence. Casein hydrolysate produced by the sequential hydrolysis of alcalase and flavourzyme showed higher degree of hydrolysis and antioxidant activity as compared to hydrolysate obtained by individual enzymes. In size exclusion chromatograph of casein hydrolysate S3, peptides with molecular weight of 0.57 kDa share 12% area in total area of chromatogram which was 10 times higher than that of hydrolysate S1 and nearly half of that of hydrolysate S2. On subjecting to HPLC-TOF-ESI separation potential antioxidant peptides were identified. The peptide sequence VLPVPQ along with potential fragments was identified in hydrolysate S1 and S2 and HPHPHLS along with its potential sequence was identified in hydrolysate S1, S2 and S3. Sequential hydrolysis of casein showed better antioxidant activity and peptide profile in less duration as compared to the casein hydrolysate obtained by individual enzyme.
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Protease from Aspergillus oryzae, ≥500 U/g
Sigma-Aldrich
Angiotensin I human acetate salt hydrate, ≥90% (HPLC), powder, suitable for mammalian cell culture
Sigma-Aldrich
Insulin Chain B Oxidized from bovine pancreas, ≥80% (HPLC), powder
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
L-Tryptophan methyl ester hydrochloride, 98%
Sigma-Aldrich
Protease from Bacillus licheniformis, ≥2.4 U/g
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)