Merck
  • Isoxazole analogues bind the system xc- transporter: structure-activity relationship and pharmacophore model.
Isoxazole analogues bind the system xc- transporter: structure-activity relationship and pharmacophore model.
Bioorganic & medicinal chemistry (2009-11-26)
Sarjubhai A Patel, Trideep Rajale, Erin O'Brien, David J Burkhart, Jared K Nelson, Brendan Twamley, Alex Blumenfeld, Monika I Szabon-Watola, John M Gerdes, Richard J Bridges, Nicholas R Natale
ABSTRACT
Analogues of amino methylisoxazole propionic acid (AMPA), were prepared from a common intermediate 12, including lipophilic analogues using lateral metalation and electrophilic quenching, and were evaluated at System xc-. Both the 5-naphthylethyl-(16) and 5-naphthylmethoxymethyl-(17) analogues adopt an E-conformation in the solid state, yet while the former has robust binding at System xc-, the latter is virtually devoid of activity. The most potent analogues were amino acid naphthyl-ACPA 7g, and hydrazone carboxylic acid, 11e Y=Y'=3,5-(CF(3))(2), which both inhibited glutamate uptake by the System xc- transporter with comparable potency to the endogenous substrate cystine, whereas in contrast the closed isoxazolo[3,4-d] pyridazinones 13 have significantly lower activity. A preliminary pharmacophore model has been constructed to provide insight into the analogue structure-activity relationships.
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%