Merck
  • SS-31 ameliorates hepatic injury in rats subjected to severe burns plus delayed resuscitation via inhibiting the mtDNA/STING pathway in Kupffer cells.
SS-31 ameliorates hepatic injury in rats subjected to severe burns plus delayed resuscitation via inhibiting the mtDNA/STING pathway in Kupffer cells.
Biochemical and biophysical research communications (2021-02-15)
Yin Wu, Chao Hao, Guangye Han, Xiongfei Liu, Changzheng Xu, Zhongtao Zou, Jinfeng Zhou, Jun Yin
ABSTRACT
Hepatic injury is common in patients who suffer from severe burns plus delayed resuscitation (B + DR). Stimulator of interferon genes (STING) is primarily expressed in Kupffer cells (KCs). We demonstrated that B + DR caused hepatic injury and oxidative stress. Reactive oxygen species (ROS) damage mitochondrial membranes in hepatocytes, leading to the release of mitochondrial DNA (mtDNA) into the hepatocyte cytosol and the circulation. The damaged hepatocytes then activate the mtDNA/STING pathway in KCs and trigger KCs polarization towards pro-inflammatory phenotype. SS-31 is a strong antioxidant that specifically concentrates in the inner mitochondrial membrane. SS-31 prevented hepatic injury by neutralizing ROS, inhibiting the release of mtDNA, protecting hepatocyte mitochondria, suppressing the activation of the mtDNA/STING pathway and inhibiting KCs polarization into pro-inflammatory phenotype.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
MISSION® esiRNA, targeting human TMEM173