Merck
  • Synthesis of some new 4(3H)-quinazolinone-2-carboxaldehyde thiosemicarbazones and their metal complexes and a study on their anticonvulsant, analgesic, cytotoxic and antimicrobial activities - part-1.
Synthesis of some new 4(3H)-quinazolinone-2-carboxaldehyde thiosemicarbazones and their metal complexes and a study on their anticonvulsant, analgesic, cytotoxic and antimicrobial activities - part-1.
European journal of medicinal chemistry (2010-06-01)
Mohsen M Aly, Yahia A Mohamed, Khairy A M El-Bayouki, Wahid M Basyouni, Samir Y Abbas
ABSTRACT
Novel 3-aryl-4(3H)-quinazolinone-2-carboxaldehydes, their corresponding Schiff's base and thiosemicarbazone derivatives were synthesized from the starting 5-iodo anthranilic acid. Copper(II), zinc(II) complexes of some thiosemicarbazone derivatives were also synthesized and characterized. Screening for some selected compounds was carried out to probe their potential anticonvulsant, analgesic, cytotoxic as well as their antimicrobial activities.
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Supelco
Norfloxacin, VETRANAL®, analytical standard
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)