Merck
  • Lithium chloride functions as Newcastle disease virus-induced ER-stress modulator and confers anti-viral effect.
Lithium chloride functions as Newcastle disease virus-induced ER-stress modulator and confers anti-viral effect.
Virus research (2020-11-10)
Kamal Shokeen, Ariktha Srivathsan, Sachin Kumar
ABSTRACT
Newcastle disease is a severe clinical manifestation of avian species caused by Newcastle disease virus (NDV). Although several vaccination strategies are available to protect poultry against NDV infection, even then, outbreaks have been reported in the vaccinated birds. The lack of therapeutics against NDV makes the need for effective anti-viral drugs is of utmost importance. Lithium Chloride (LiCl) is a widely prescribed drug for the treatment of bipolar disorder, acute brain injuries, and chronic neurodegenerative diseases. Also, LiCl has been repurposed as an effective anti-viral drug for some viral infections. In the present work, we have investigated the efficacy of LiCl to inhibit NDV replication using in vitro, in ovo, and in vivo models. Our results collectively showed the modulation of NDV replication after the LiCl treatment. We also demonstrated that NDV induces endoplasmic reticulum stress (ER-stress), and a stress-inducible ER chaperone, glucose-regulating protein 78 (GRP78), was found to be over-expressed after NDV infection. Subsequently, the treatment of NDV infected cells with LiCl significantly reduced the transcript and protein levels of GRP78. Finally, we concluded that LiCl treatment protects the cells from ER-stress induced by the NDV infection.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis