Merck
  • Interplay between drug efflux and antioxidants in Escherichia coli resistance to antibiotics.
Interplay between drug efflux and antioxidants in Escherichia coli resistance to antibiotics.
Antimicrobial agents and chemotherapy (2010-09-30)
Girija Dhamdhere, Ganesh Krishnamoorthy, Helen I Zgurskaya
ABSTRACT
We investigated possible cross talk between endogenous antioxidants glutathione, spermidine, and glutathionylspermidine and drug efflux in Escherichia coli. We found that cells lacking either spermidine or glutathione are less susceptible than the wild type to novobiocin and certain aminoglycosides. In contrast, exogenous glutathione protects against both bactericidal and bacteriostatic antibiotics. The glutathione protection does not require the AcrAB efflux pump but fails in cells lacking TolC because exogenous glutathione is toxic to these cells.
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.