Merck
  • Phenolic constituents of the aerial parts of Cimicifuga simplex and Cimicifuga japonica.
Phenolic constituents of the aerial parts of Cimicifuga simplex and Cimicifuga japonica.
Journal of natural products (2010-02-27)
Atsufumi Iwanaga, Genjiro Kusano, Tsutomu Warashina, Toshio Miyase
ABSTRACT
Chemical investigation of the aerial parts of Cimicifuga simplex afforded four new fukinolic acid analogues, cimicifugic acids K-N (1-4), and 10 known compounds, and C. japonica afforded three new fukinolic acid analogues, cimicifugic acids K-M (1-3), a new phenolic glycoside, shomaside F (5), and 10 known compounds. Cimicifugic acids K-N showed more potent hyaluronidase inhibitory activities than rosmarinic acid.
Sigma-Aldrich
Rosmarinic acid, 96%
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3-Hydroxy-4-methoxycinnamic acid, predominantly trans, 97%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)