Merck
  • Mammalian lipid droplets are innate immune hubs integrating cell metabolism and host defense.
Mammalian lipid droplets are innate immune hubs integrating cell metabolism and host defense.
Science (New York, N.Y.) (2020-10-17)
Marta Bosch, Miguel Sánchez-Álvarez, Alba Fajardo, Ronan Kapetanovic, Bernhard Steiner, Filipe Dutra, Luciana Moreira, Juan Antonio López, Rocío Campo, Montserrat Marí, Frederic Morales-Paytuví, Olivia Tort, Albert Gubern, Rachel M Templin, James E B Curson, Nick Martel, Cristina Català, Francisco Lozano, Francesc Tebar, Carlos Enrich, Jesús Vázquez, Miguel A Del Pozo, Matthew J Sweet, Patricia T Bozza, Steven P Gross, Robert G Parton, Albert Pol
ABSTRACT
Lipid droplets (LDs) are the major lipid storage organelles of eukaryotic cells and a source of nutrients for intracellular pathogens. We demonstrate that mammalian LDs are endowed with a protein-mediated antimicrobial capacity, which is up-regulated by danger signals. In response to lipopolysaccharide (LPS), multiple host defense proteins, including interferon-inducible guanosine triphosphatases and the antimicrobial cathelicidin, assemble into complex clusters on LDs. LPS additionally promotes the physical and functional uncoupling of LDs from mitochondria, reducing fatty acid metabolism while increasing LD-bacterial contacts. Thus, LDs actively participate in mammalian innate immunity at two levels: They are both cell-autonomous organelles that organize and use immune proteins to kill intracellular pathogens as well as central players in the local and systemic metabolic adaptation to infection.
Sigma-Aldrich
Ketone Body Assay, sufficient for 200 tests (UV)
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
Millipore
Protein G Sepharose, Fast Flow, recombinant, expressed in E. coli, aqueous ethanol suspension
Sigma-Aldrich
Triton X-100, Molecular Biology
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)