Merck
  • Serotonin 2B Receptor by Interacting with NMDA Receptor and CIPP Protein Complex May Control Structural Plasticity at Glutamatergic Synapses.
Serotonin 2B Receptor by Interacting with NMDA Receptor and CIPP Protein Complex May Control Structural Plasticity at Glutamatergic Synapses.
ACS chemical neuroscience (2021-03-20)
Amina Benhadda, Emily Quentin, Imane Moutkine, Benjamin Chanrion, Marion Russeau, Philippe Marin, Sabine Levi, Luc Maroteaux
ABSTRACT
The serotonin 2B (5-HT2B) receptor coupled to Gq-protein contributes to the control of neuronal excitability and is implicated in various psychiatric disorders. The mechanisms underlying its brain function are not fully described. Using peptide affinity chromatography combined with mass spectrometry, we found that the PDZ binding motif of the 5-HT2B receptor located at its C-terminal end interacts with the scaffolding protein channel interacting PDZ protein (CIPP). We then showed, in COS-7 cells, that the association of the 5-HT2B receptor to CIPP enhanced receptor-operated inositol phosphate (IP) production without affecting its cell surface and intracellular levels. Co-immunoprecipitation experiments revealed that CIPP, the 5-HT2B receptor, and the NR1 subunit of the NMDA receptor form a macromolecular complex. CIPP increased 5-HT2B receptor clustering at the surface of primary cultured hippocampal neurons and prevented receptor dispersion following agonist stimulation, thus potentiating IP production and intracellular calcium mobilization in dendrites. CIPP or 5-HT2B receptor stimulation in turn dispersed NR1 clusters colocalized with 5-HT2B receptors and increased the density and maturation of dendritic spines. Collectively, our results suggest that the 5-HT2B receptor, the NMDA receptor, and CIPP may form a signaling platform by which serotonin can influence structural plasticity of excitatory glutamatergic synapses.
Sigma-Aldrich
Anti-Neurofilament 200 kDa Antibody, clone RT97, clone RT97, Chemicon®, from mouse
Sigma-Aldrich
Anti-NMDAR1 Antibody, clone 54.1, clone 54.1, Chemicon®, from mouse
Supelco
Serotonin, analytical standard
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid