Merck
  • Three-dimensional model of glioblastoma by co-culturing tumor stem cells with human brain organoids.
Three-dimensional model of glioblastoma by co-culturing tumor stem cells with human brain organoids.
Biology open (2021-02-24)
Roberta Azzarelli, Michela Ori, Anna Philpott, Benjamin D Simons
ABSTRACT
Emerging three-dimensional (3D) cultures of glioblastoma are becoming powerful models to study glioblastoma stem cell behavior and the impact of cell-cell and cell-microenvironment interactions on tumor growth and invasion. Here we describe a method for culturing human glioblastoma stem cells (GSCs) in 3D by co-culturing them with pluripotent stem cell-derived brain organoids. This requires multiple coordinated steps, including the generation of cerebral organoids, and the growth and fluorescence tagging of GSCs. We highlight how to recognize optimal organoid generation and how to efficiently mark GSCs, before describing optimized co-culture conditions. We show that GSCs can efficiently integrate into brain organoids and maintain a significant degree of cell fate heterogeneity, paving the way for the analysis of GSC fate behavior and lineage progression. These results establish the 3D culture system as a viable and versatile GBM model for investigating tumor cell biology and GSC heterogeneity.This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Hexadimethrine bromide, ≥94% (titration)
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Donkey serum
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture