Merck
  • Crassocolides N-P, three cembranoids from the Formosan soft coral Sarcophyton crassocaule.
Crassocolides N-P, three cembranoids from the Formosan soft coral Sarcophyton crassocaule.
Bioorganic & medicinal chemistry letters (2011-10-22)
Guey-Horng Wang, Ho-Cheng Huang, Jui-Hsin Su, Chiung-Yao Huang, Chi-Hsin Hsu, Yao-Haur Kuo, Jyh-Horng Sheu
ABSTRACT
Three new cembranoids crassocolides N-P (1-3), was isolated from the organic extract of a Formosan soft coral Sarcophyton crassocaule. These structures were elucidated on the basis of spectroscopic analyses and by comparison with those previously reported in literature. The cytotoxicity of these compounds toward various cancer cell lines has also been determined.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture