Merck
  • Predicting hERG activities of compounds from their 3D structures: development and evaluation of a global descriptors based QSAR model.
Predicting hERG activities of compounds from their 3D structures: development and evaluation of a global descriptors based QSAR model.
European journal of medicinal chemistry (2010-12-28)
Nandita Sinha, Srikanta Sen
ABSTRACT
A QSAR based predictive model of hERG activity in terms of 'global descriptors' has been developed and evaluated. The QSAR was developed by training 77 compounds covering a wide range of activities and was validated based on an external 'test set' of 80 compounds using neural network method. Statistical parameters and examination of enrichment factor indicated the effectiveness of the present model. Randomization test demonstrated the robustness of the model and cross-validation test further validated the QSAR. Domain of applicability test indicated to the high degree of reliability of the predicted results. Satisfactory performance in classifying compounds into 'active' and 'inactive' groups was also obtained. The cases where the QSAR failed, the possible sources of errors have been discussed.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
4-Aminopyridine, 98%
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Carbamazepine, powder
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Trimethoprim, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Nitrendipine, >95%, powder
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
N-Desmethylclozapine
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration), meets EP testing specifications
Sigma-Aldrich
Terfenadine