Merck
  • Synthesis of new 5,6-dihydrobenzo[h]quinazoline 2,4-diamino substituted and antiplatelet/antiphlogistic activities evaluation.
Synthesis of new 5,6-dihydrobenzo[h]quinazoline 2,4-diamino substituted and antiplatelet/antiphlogistic activities evaluation.
Bioorganic & medicinal chemistry letters (2011-12-23)
Chiara Brullo, Massimo Rocca, Paola Fossa, Elena Cichero, Elisabetta Barocelli, Vigilio Ballabeni, Lisa Flammini, Carmine Giorgio, Francesca Saccani, Giuseppe Domenichini, Olga Bruno
ABSTRACT
In pursuing our research on some 2,4-diamino-benzopyranopyrimidines and 2-amino-5,6-dihydrobenzo[h]quinazolines, previously reported as antiplatelet and analgesic/anti-inflammatory agents respectively, we designed and synthesized a new series of 5,6-dihydrobenzo[h]quinazoline 2,4-diamino substituted. The insertion of amino substituents at positions 2 and 4 of the benzoquinazoline scaffold resulted in compounds endowed with a potent ASA-like antiplatelet activity, combined with an anti-inflammatory activity comparable, in some cases, to that of indomethacin, used as a reference drug.
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications