Merck
  • Liquid culture system for efficient depletion of the endogenous nutrients in Arabidopsis seedlings.
Liquid culture system for efficient depletion of the endogenous nutrients in Arabidopsis seedlings.
STAR protocols (2021-11-12)
Yanlin Liu, Yan Xiong
ABSTRACT
Target of rapamycin (TOR) is a central regulator in nutrient signaling. However, the innate capacity of autotrophic plants to produce carbon-related nutrients and nitrogen-related nutrients makes studying the TOR pathway difficult. Here, we describe a protocol for a liquid culture system for efficient depletion of the endogenous carbon/nitrogen nutrients in Arabidopsis seedlings. Exogenous carbon/nitrogen can be supplied to dissect the TOR pathway. For complete details on the use and execution of this protocol, please refer to Xiong et al. (2013) and Liu et al. (2021).
Product Description
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
5-Bromo-4-chloro-3-indolyl β-D-glucuronide sodium salt, ≥98%
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, ACS reagent, ≥99.0%