Merck
  • Effects of Pulsed Electric Field Processing and Sous Vide Cooking on Muscle Structure and In Vitro Protein Digestibility of Beef Brisket.
Effects of Pulsed Electric Field Processing and Sous Vide Cooking on Muscle Structure and In Vitro Protein Digestibility of Beef Brisket.
Foods (Basel, Switzerland) (2021-04-04)
Feng Ming Chian, Lovedeep Kaur, Indrawati Oey, Thierry Astruc, Suzanne Hodgkinson, Mike Boland
ABSTRACT
Pulsed electric fields (PEF) in conjunction with sous vide (SV) cooking has been explored for meat tenderisation. The aim of this experiment was to study the effect of PEF-SV treatment on the muscle structure and in vitro protein digestibility of beef brisket. Pulsed electric field treatment (specific energy of 99 ± 5 kJ/kg) was applied to bovine Deep and Superficial pectoral muscles in combination with sous vide (SV) cooking (60 °C for 24 h). A similar micro- and ultrastructure was detected between the control SV-cooked and PEF-treated SV-cooked pectoral muscles. The combined PEF-SV treatment increased the in vitro protein digestibility of the pectoral muscles by approximately 29%, in terms of ninhydrin-reactive free amino nitrogen released at the end of simulated digestion. An increment in proteolysis of the PEF-treated SV-cooked meat proteins (e.g., myosin heavy chains and C-protein) during simulated digestion was also observed using sodium dodecyl sulfate-polyacrylamide gel electrophoresis. More damaged muscle micro- and ultrastructure was detected in PEF-treated SV-cooked muscles at the end of in vitro digestion, showing its enhanced digestive proteolysis compared to the control cooked meat.
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use
Sigma-Aldrich
Ninhydrin Reagent, 2% Solution
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein