Merck
  • Synthesis of Recombinant Human Hemoglobin With NH2 -Terminal Acetylation in Escherichia coli.
Synthesis of Recombinant Human Hemoglobin With NH2 -Terminal Acetylation in Escherichia coli.
Current protocols in protein science (2020-07-21)
Chandrasekhar Natarajan, Anthony V Signore, Vikas Kumar, Jay F Storz
ABSTRACT
The development of new technologies for the efficient expression of recombinant hemoglobin (rHb) is of interest for experimental studies of protein biochemistry and the development of cell-free blood substitutes in transfusion medicine. Expression of rHb in Escherichia coli host cells has numerous advantages, but one disadvantage of using prokaryotic systems to express eukaryotic proteins is that they are incapable of performing post-translational modifications such as NH2 -terminal acetylation. One possible solution is to coexpress additional enzymes that can perform the necessary modifications in the host cells. Here, we report a new method for synthesizing human rHb with proper NH2 -terminal acetylation. Mass spectrometry experiments involving native and recombinant human Hb confirmed the efficacy of the new technique in producing correctly acetylated globin chains. Finally, functional experiments provided insights into the effects of NH2 -terminal acetylation on O2 binding properties. © 2020 Wiley Periodicals LLC. Basic Protocol 1: Gene synthesis and cloning the cassette to the expression plasmid Basic Protocol 2: Selection of E. coli expression strains for coexpression Basic Protocol 3: Large-scale recombinant hemoglobin expression and purification Support Protocol 1: Measuring O2 equilibration curves Support Protocol 2: Mass spectrometry to confirm NH2 -terminal acetylation.
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Kanamycin sulfate, mixture of Kanamycin A (main component) and Kanamycin B and C
Sigma-Aldrich
LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trizma® hydrochloride, reagent grade, ≥99.0% (titration), crystalline
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, Molecular Biology, suitable for electrophoresis, ≥98%
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Sodium hydrosulfite, technical grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%