Merck
  • Screening of novel Midkine binding protein by BioID2-based proximity labeling.
Screening of novel Midkine binding protein by BioID2-based proximity labeling.
Nagoya journal of medical science (2021-09-24)
Yosuke Komata, Shoma Tsubota, Kazuma Sakamoto, Shinya Ikematsu, Kenji Kadomatsu
ABSTRACT
Midkine (MK), a heparin-binding growth factor, is associated with the poor prognosis of the pediatric tumor, neuroblastoma. MK would be a druggable target as many studies showed inhibition of its function in various cancers suppressed tumor developments. To establish the therapy targeting MK, identification of its binding partners, and elucidation of its intracellular signaling are needed. It was reported that exogenous MK induced phosphorylation of ribosomal protein S6 (RPS6) downstream of mTOR signaling. Using RPS6 phosphorylation as a marker of MK response, we searched for MK reactive cell lines. We found that MK cell lines expressing less MK tended to respond better to MK. Next, using an MK reactive neuroblastoma cell line, MK-knocked down SH-SY5Y cells, we employed a proximity-dependent biotin identification method, which was invented to evaluate protein-protein interactions by biotinylation. We confirmed that secreted MK fused to the biotin ligase BioID2 (MK-BioID2) was able to biotinylate proteins from the cells. Biotinylated proteins were identified by liquid chromatography-mass spectrometry analyses. Twenty five proteins were found to be overlapped after three independent experiments, among which insulin-like growth binding protein 2 (IGFBP2) was further analyzed. IGFBP2 was indeed detected with immunoblotting after streptavidin pull down of MK-BioID2 labeled cell extract of MK-knocked down SH-SY5Y cells. Our study suggests that the BioID2 method is useful to identify binding partners of growth factors.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Collagen Type I, rat tail, 100 mg, solution, BioReagent, suitable for cell culture