Merck
  • (Phospho)Proteomic dataset of ischemia- and ultrasound- stimulated mouse cardiac endothelial cells in vitro.
(Phospho)Proteomic dataset of ischemia- and ultrasound- stimulated mouse cardiac endothelial cells in vitro.
Data in brief (2021-09-17)
Uchenna Emechebe, David Giraud, Azzdine Y Ammi, Kristin L Scott, Jon M Jacobs, Jason E McDermott, Igor V Dykan, Nabil J Alkayed, Anthony P Barnes, Sanjiv Kaul, Catherine M Davis
ABSTRACT
Cardiac endothelial cells respond to both ischemia and therapeutic ultrasound; the proteomic changes underlying these responses are unknown. This data article provides raw and processed data resulting from our global, unbiased phosphoproteomics investigation conducted on primary mouse cardiac endothelial cells exposed to ischemia (2-hour oxygen glucose deprivation) and ultrasound (250 kHz, 1.2 MPa) in vitro [1]. Proteins were extracted from cell lysates and enriched phosphopeptides were analyzed with a high mass accuracy liquid chromatrography (LC) - tandem mass spectrometry (MS/MS) proteomic platform, yielding multiple alterations in both total protein levels and phosphorylation events in response to ischemic injury and ultrasound. This dataset can be used as a reference for future studies on the cardiac endothelial response to ischemia and the mechanistic underpinnings of the cellular response to ultrasound, with the potential to yield clinically relevant therapeutic targets.
Product Description
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Gentamicin solution, 50 mg/mL in deionized water, liquid, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, USA origin, suitable for cell culture
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)