Merck
In Vitro Cultivation of the Syphilis Spirochete Treponema pallidum.
Current protocols (2021-02-19)
Diane G Edmondson, Steven J Norris
ABSTRACT
For over a century, investigation of Treponema pallidum subsp. pallidum, the spiral-shaped bacterium that causes syphilis, was hindered by an inability to culture the organism in vitro. A recent breakthrough has enabled continuous in vitro growth of this organism in co-culture with mammalian tissue culture cells. This article contains the protocols needed to culture T. pallidum in the standard laboratory environment. In addition, protocols for growing and maintaining the required tissue culture cells, for generating isogenic strains by limiting dilution, and for quantitating T. pallidum by darkfield microscopy are included. © 2021 Wiley Periodicals LLC. Basic Protocol 1: In vitro cultivation of Treponema pallidum Basic Protocol 2: Generation of isogenic strains Support Protocol 1: Alternate harvest procedure Support Protocol 2: Culture of Sf1Ep cells Support Protocol 3: Assessment of T. pallidum number and viability.
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Resazurin sodium salt, powder, BioReagent
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%