Merck
  • Development of a novel in vitro assay to screen for neuroprotective drugs against iatrogenic neurite shortening.
Development of a novel in vitro assay to screen for neuroprotective drugs against iatrogenic neurite shortening.
PloS one (2021-03-11)
Antón L Martínez, José Brea, Mateo Barro, Xavier Monroy, Manuel Merlos, Javier Burgueño, María Isabel Loza
ABSTRACT
This work tries to help overcome the lack of relevant translational screening assays, as a limitation for the identification of novel analgesics for neuropathic pain. Hyperexcitability and neurite shortening are common adverse effects of antiviral and antitumor drugs, leading to neuropathic pain. Now, as seen in the drug screening that we developed here, a high-content microscopy-based assay with immortalized dorsal root ganglia (DRG) neurons (differentiated F11 cells) allowed to identify drugs able to protect against the iatrogenic neurite shortening induced by the antitumor drug vincristine and the antiviral drug rilpivirine. We observed that vincristine and rilpivirine induced a significant reduction in the neurite length, which was reverted by α-lipoic acid. We had also evidenced protective effects of pregabalin and melatonin, acting through the α2δ-2 subunit of the voltage-dependent calcium channels and the MT1 receptor, respectively. Additionally, two hits originated from a previous primary screening aimed to detect inhibitors of hyperexcitability to inflammatory mediators in DRG neurons (nitrendipine and felodipine) also prevented neurite shortening in our model. In summary, in this work we developed a novel secondary assay for identifying hits with neuroprotective effect against iatrogenic neurite shortening, consistent with the anti-hyperexcitability action previously tested: highlighting nitrendipine and felodipine against iatrogenic damage in DRG neurons.
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
(±)-α-Lipoic acid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Triton X-100, Molecular Biology
Corning® Costar® TC-Treated Multiple Well Plates, size 6 wells, clear, polystyrene plate, flat bottom, case of 50 (individually wrapped), sterile, lid
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine and sodium pyruvate, liquid, sterile-filtered, suitable for cell culture, suitable for hybridoma
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Roche
Bovine Serum Albumin, 98.5% (electrophoresis), Fatty acids (total) ≤0.2 mg/g, Triglycerides (enzym.) free, Immunoglobulins not detectable, microbiological culture: suitable, USA origin