Merck
  • Altered synaptic glutamate homeostasis contributes to cognitive decline in young APP/PSEN1 mice.
Altered synaptic glutamate homeostasis contributes to cognitive decline in young APP/PSEN1 mice.
Neurobiology of disease (2021-08-28)
J M Wilcox, D C Consoli, A A Tienda, S Dixit, R A Buchanan, J M May, W P Nobis, F E Harrison
ABSTRACT
Non-convulsive epileptiform activity is a common and under-studied comorbidity of Alzheimer's disease that may significantly contribute to onset of clinical symptoms independently of other neuropathological features such as β-amyloid deposition. We used repeated treatment with low dose kainic acid (KA) to trigger sub-threshold epileptiform activity in young (less than 6 months) wild-type (WT) and APP/PSEN1 mice to test the role of disruption to the glutamatergic system in epileptiform activity changes and the development of memory deficits. Short-term repeated low-dose KA (five daily treatments with 5 mg/kg, IP) impaired long-term potentiation in hippocampus of APP/PSEN1 but not WT mice. Long-term repeated low-dose KA (fourteen weeks of bi-weekly treatment with 7.5-10 mg/kg) led to high mortality in APP/PSEN1 mice. KA treatment also impaired memory retention in the APP/PSEN1 mice in a Morris water maze task under cognitively challenging reversal learning conditions where the platform was moved to a new location. Four weeks of bi-weekly treatment with 5 mg/kg KA also increased abnormal spike activity in APP/PSEN1 and not WT mice but did not impact sleep/wake behavioral states. These findings suggest that hyperexcitability in Alzheimer's disease may indeed be an early contributor to cognitive decline that is independent of heavy β-amyloid-plaque load, which is absent in APP/PSEN1 mice under 6 months of age.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, ascites fluid, clone GA5, Chemicon®
Sigma-Aldrich
Anti-Glutamate Transporter Antibody, Glial, serum, Chemicon®
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Kainic acid monohydrate, ≥99% (TLC)