Merck
  • α-Synuclein pre-formed fibrils impair tight junction protein expression without affecting cerebral endothelial cell function.
α-Synuclein pre-formed fibrils impair tight junction protein expression without affecting cerebral endothelial cell function.
Experimental neurology (2016-09-17)
Wei-Li Kuan, Neal Bennett, Xiaoling He, Jeremy N Skepper, Nataly Martynyuk, Ruwani Wijeyekoon, Prabhas V Moghe, Caroline H Williams-Gray, Roger A Barker
ABSTRACT
Recently it has been shown that there is impaired cerebral endothelial function in many chronic neurodegenerative disorders including Alzheimer's and Huntington's disease. Such problems have also been reported in Parkinson's disease, in which α-synuclein aggregation is the pathological hallmark. However, little is known about the relationship between misfolded α-synuclein and endothelial function. In the present study, we therefore examined whether α-synuclein preformed fibrils affect endothelial function in vitro. Using a well-established endothelial cell model, we found that the expression of tight junction proteins, in particular zona occludens-1 and occludin, was significantly perturbed in the presence of fibril-seeded neurotoxicity. Disrupted expression of these proteins was also found in the postmortem brains of patients dying with Parkinson's disease. There was though little evidence in vitro of functional impairments in endothelial cell function in terms of transendothelial electrical resistance and permeability. This study therefore shows for the first time that misfolded α-synuclein can interact and affect the cerebral endothelial system, although its relevance to the pathogenesis of Parkinson's disease remains to be elucidated.
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 10,000
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt ≥300,000, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks