- Utilizing thermal proteome profiling to identify the molecular targets of anti-leishmanial compounds.
Utilizing thermal proteome profiling to identify the molecular targets of anti-leishmanial compounds.
STAR protocols (2021-09-02)
Victoriano Corpas-Lopez, Susan Wyllie
PMID34467225
ABSTRACT
Here, we detail our optimized protocol for the identification of drug targets in Leishmania donovani using thermal proteome profiling. This approach is based on the principle that binding of a drug to its protein target can significantly alter the thermal stability of that protein. By monitoring changes in the thermal stability of proteins within drug-treated and untreated cell lysates, using mass spectrometry combined with tandem mass tag labeling, putative targets of the drug can be identified in an unbiased manner. For further details on the use and application of this protocol, please refer to Paradela et al. (2021).
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Medium 199, With Hanks′ salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Roche
cOmplete™, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial