Merck
  • Pork Liver Pâté Enriched with Persimmon Coproducts: Effect of In Vitro Gastrointestinal Digestion on Its Fatty Acid and Polyphenol Profile Stability.
Pork Liver Pâté Enriched with Persimmon Coproducts: Effect of In Vitro Gastrointestinal Digestion on Its Fatty Acid and Polyphenol Profile Stability.
Nutrients (2021-05-01)
Raquel Lucas-González, José Ángel Pérez-Álvarez, Manuel Viuda-Martos, Juana Fernández-López
ABSTRACT
Agrofood coproducts are used to enrich meat products to reduce harmful compounds and contribute to fiber and polyphenol enrichment. Pork liver pâtés with added persimmon coproducts (3 and 6%; PR-3 and PR-6, respectively) were developed. Therefore, the aim was to study the effect of their in vitro gastrointestinal digestion on: the free and bound polyphenol profile (HPLC) and their colon-available index; the lipid oxidation (TBARs); and the stability of the fatty acid profile (GC). Furthermore, the effect of lipolysis was investigated using two pancreatins with different lipase activity. Forty-two polyphenols were detected in persimmon flour, which were revealed as a good source of bound polyphenols in pâtés, especially gallic acid (164.3 µg/g d.w. in PR-3 and 631.8 µg/g d.w. in PR-6). After gastrointestinal digestion, the colon-available index in enriched pâté ranged from 88.73 to 195.78%. The different lipase activity in the intestinal phase caused significant differences in bound polyphenols' stability, contributing to increased lipid oxidation. The fatty acids profile in pâté samples was stable, and surprisingly their PUFA content was raised. In conclusion, rich fatty foods, such as pâté, are excellent vehicles to preserve bound polyphenols, which can reach the colon intact and be metabolized by the intestinal microbiome.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Bile from bovine and ovine, bile acid mixture
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications