Merck
Soft artificial electroreceptors for noncontact spatial perception.
Science advances (2021-11-25)
Won Jun Song, Younghoon Lee, Yeonsu Jung, Yong-Woo Kang, Junhyung Kim, Jae-Man Park, Yong-Lae Park, Ho-Young Kim, Jeong-Yun Sun
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Lithium phenyl-2,4,6-trimethylbenzoylphosphinate, ≥95%