Merck
  • Analysis of the proteolysis of bioactive peptides using a peptidomics approach.
Analysis of the proteolysis of bioactive peptides using a peptidomics approach.
Nature protocols (2013-08-21)
Yun-Gon Kim, Anna Mari Lone, Alan Saghatelian
ABSTRACT
Identifying the peptidases that inactivate bioactive peptides (e.g., peptide hormones and neuropeptides) in mammals is an important unmet challenge. This protocol describes a recent approach that uses liquid chromatography-mass spectrometry (LC-MS) peptidomics to identify endogenous cleavage sites of a bioactive peptide; it also addresses the subsequent biochemical purification of a candidate peptidase on the basis of these cleavage sites and the validation of the candidate peptidase's role in the physiological regulation of the bioactive peptide by examining a peptidase-knockout mouse. We highlight the successful application of this protocol in the discovery that insulin-degrading enzyme (IDE) regulates physiological calcitonin gene-related peptide (CGRP) levels, and we detail the key stages and steps in this approach. This protocol requires 7 d of work; however, the total time for this protocol is highly variable because of its dependence on the availability of biological reagents such as purified enzymes and knockout mice. The protocol is valuable because it expedites the characterization of mammalian peptidases, such as IDE, which in certain instances can be used to develop novel therapeutics.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Bestatin hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
1,10-Phenanthroline monohydrate, reagent grade