Merck
  • Water-mediated conformations of the alanine dipeptide as revealed by distributed umbrella sampling simulations, quantum mechanics based calculations, and experimental data.
Water-mediated conformations of the alanine dipeptide as revealed by distributed umbrella sampling simulations, quantum mechanics based calculations, and experimental data.
The journal of physical chemistry. B (2011-04-08)
Víctor Cruz, Javier Ramos, Javier Martínez-Salazar
ABSTRACT
An exhaustive umbrella sampling simulation of the alanine dipeptide in solution is reported. The presence of stable Y conformations in solution is assessed by both quantum calculations and experimental data from X-ray and NMR protein-solved structures available in the protein coil library. The agreement between experimental and simulation Ramachandran plots of the dipeptide in solution is excellent. A suitable explanation of the stabilization of the Y conformation mediated by water for the alanine dipeptide is discussed on the basis of Car-Parrinello MD calculations of the dipeptide in water.
Sigma-Aldrich
Ala-Gly
Sigma-Aldrich
Ala-Phe
Sigma-Aldrich
Ala-Leu
Sigma-Aldrich
Ala-Tyr
Sigma-Aldrich
Ala-Pro hydrate