Merck
  • Yeast adaptive response to acetic acid stress involves structural alterations and increased stiffness of the cell wall.
Yeast adaptive response to acetic acid stress involves structural alterations and increased stiffness of the cell wall.
Scientific reports (2021-06-18)
Ricardo A Ribeiro, Miguel V Vitorino, Cláudia P Godinho, Nuno Bourbon-Melo, Tiago T Robalo, Fábio Fernandes, Mário S Rodrigues, Isabel Sá-Correia
ABSTRACT
This work describes a coordinate and comprehensive view on the time course of the alterations occurring at the level of the cell wall during adaptation of a yeast cell population to sudden exposure to a sub-lethal stress induced by acetic acid. Acetic acid is a major inhibitory compound in industrial bioprocesses and a widely used preservative in foods and beverages. Results indicate that yeast cell wall resistance to lyticase activity increases during acetic acid-induced growth latency, corresponding to yeast population adaptation to sudden exposure to this stress. This response correlates with: (i) increased cell stiffness, assessed by atomic force microscopy (AFM); (ii) increased content of cell wall β-glucans, assessed by fluorescence microscopy, and (iii) slight increase of the transcription level of the GAS1 gene encoding a β-1,3-glucanosyltransferase that leads to elongation of (1→3)-β-D-glucan chains. Collectively, results reinforce the notion that the adaptive yeast response to acetic acid stress involves a coordinate alteration of the cell wall at the biophysical and molecular levels. These alterations guarantee a robust adaptive response essential to limit the futile cycle associated to the re-entry of the toxic acid form after the active expulsion of acetate from the cell interior.
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥2,000 units/mg protein, Protein ≥20 % by biuret
Sigma-Aldrich
Lyticase from Arthrobacter luteus, lyophilized powder, ≥200 units/mg solid