Merck
  • A distinct glycoform of CD44 is an L-selectin ligand on human hematopoietic cells.
A distinct glycoform of CD44 is an L-selectin ligand on human hematopoietic cells.
Proceedings of the National Academy of Sciences of the United States of America (2000-11-30)
C J Dimitroff, J Y Lee, R C Fuhlbrigge, R Sackstein
ABSTRACT
We previously have obtained operational evidence of a hematopoietic cell L-selectin ligand expressed on normal human hematopoietic cells and on leukemic blasts. Using a technique developed in our laboratory for analyzing and identifying adhesion molecules, we show here that hematopoietic cell L-selectin ligand is a specialized glycoform of CD44. This L-selectin ligand activity of CD44 requires sialofucosylated N-linked glycans and is sulfation-independent. These data provide important insights on the structural biology of CD44 and reveal a role for this protein as an L-selectin ligand on human hematopoietic cells.