Merck
  • TIFA has dual functions in Helicobacter pylori-induced classical and alternative NF-κB pathways.
TIFA has dual functions in Helicobacter pylori-induced classical and alternative NF-κB pathways.
EMBO reports (2021-07-31)
Gunter Maubach, Michelle C C Lim, Olga Sokolova, Steffen Backert, Thomas F Meyer, Michael Naumann
ABSTRACT
Helicobacter pylori infection constitutes one of the major risk factors for the development of gastric diseases including gastric cancer. The activation of nuclear factor-kappa-light-chain-enhancer of activated B cells (NF-κB) via classical and alternative pathways is a hallmark of H. pylori infection leading to inflammation in gastric epithelial cells. Tumor necrosis factor receptor-associated factor (TRAF)-interacting protein with forkhead-associated domain (TIFA) was previously suggested to trigger classical NF-κB activation, but its role in alternative NF-κB activation remains unexplored. Here, we identify TRAF6 and TRAF2 as binding partners of TIFA, contributing to the formation of TIFAsomes upon H. pylori infection. Importantly, the TIFA/TRAF6 interaction enables binding of TGFβ-activated kinase 1 (TAK1), leading to the activation of classical NF-κB signaling, while the TIFA/TRAF2 interaction causes the transient displacement of cellular inhibitor of apoptosis 1 (cIAP1) from TRAF2, and proteasomal degradation of cIAP1, to facilitate the activation of the alternative NF-κB pathway. Our findings therefore establish a dual function of TIFA in the activation of classical and alternative NF-κB signaling in H. pylori-infected gastric epithelial cells.
Sigma-Aldrich
Dynamin Inhibitor I, Dynasore, The Dynamin Inhibitor I, Dynasore, also referenced under CAS 304448-55-3, controls the biological activity of Dynamin. This small molecule/inhibitor is primarily used for Membrane applications.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)