Merck
  • Printing biohybrid materials for bioelectronic cardio-3D-cellular constructs.
Printing biohybrid materials for bioelectronic cardio-3D-cellular constructs.
iScience (2022-07-06)
Paola Sanjuan-Alberte, Charlie Whitehead, Joshua N Jones, João C Silva, Nathan Carter, Simon Kellaway, Richard J M Hague, Joaquim M S Cabral, Frederico C Ferreira, Lisa J White, Frankie J Rawson
ABSTRACT
Conductive hydrogels are emerging as promising materials for bioelectronic applications as they minimize the mismatch between biological and electronic systems. We propose a strategy to bioprint biohybrid conductive bioinks based on decellularized extracellular matrix (dECM) and multiwalled carbon nanotubes. These inks contained conductive features and morphology of the dECM fibers. Electrical stimulation (ES) was applied to bioprinted structures containing human pluripotent stem cell-derived cardiomyocytes (hPSC-CMs). It was observed that in the absence of external ES, the conductive properties of the materials can improve the contractile behavior of the hPSC-CMs, and this effect is enhanced under the application of external ES. Genetic markers indicated a trend toward a more mature state of the cells with upregulated calcium handling proteins and downregulation of calcium channels involved in the generation of pacemaking currents. These results demonstrate the potential of our strategy to manufacture conductive hydrogels in complex geometries for actuating purposes.
Product Description
Roche
Papain, from Carica papaya
Sigma-Aldrich
Monoclonal Anti-TNNI3 antibody produced in mouse, clone 1E7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥2,500 units/mg protein (E1%/280)