Merck
  • The BAX-binding protein MOAP1 associates with LC3 and promotes closure of the phagophore.
The BAX-binding protein MOAP1 associates with LC3 and promotes closure of the phagophore.
Autophagy (2021-03-31)
Hao-Chun Chang, Ran N Tao, Chong Teik Tan, Ya Jun Wu, Boon Huat Bay, Victor C Yu
ABSTRACT
MOAP1 (modulator of apoptosis 1) is a BAX-binding protein tightly regulated by the ubiquitin-proteasome system. Apoptotic stimuli stabilize MOAP1 protein and facilitate its interaction with BAX to promote apoptosis. Here we show that in contrast to being resistant to apoptotic stimuli, MOAP1-deficient cells are hypersensitive to cell death mediated by starvation rendered by EBSS treatment. MOAP1-deficient cells exhibited impairment in macroautophagy/autophagy signaling induced by EBSS. Mechanistic analysis revealed that MOAP1-deficient cells had no notable defect in the recruitment of the pre-autophagosomal phosphatidylinositol-3-phosphate (PtdIns3P)-binding proteins, ZFYVE1/DFCP1 and WIPI2, nor in the LC3 lipidation mechanism regulated by the ATG12-ATG5-ATG16L1 complex upon EBSS treatment. Interestingly, MOAP1 is required for facilitating efficient closure of phagophore in the EBSS-treated cells. Analysis of LC3-positive membrane structures using Halo-tagged LC3 autophagosome completion assay showed that predominantly unclosed phagophore rather than closed autophagosome was present in the EBSS-treated MOAP1-deficient cells. The autophagy substrate SQSTM1/p62, which is normally contained within the enclosed autophagosome under EBSS condition, was also highly sensitive to degradation by proteinase K in the absence of MOAP1. MOAP1 binds LC3 and the binding is critically dependent on a LC3-interacting region (LIR) motif detected at its N-terminal region. Re-expression of MOAP1, but not its LC3-binding defective mutant, MOAP1-LIR, in the MOAP1-deficient cells, restored EBSS-induced autophagy. Together, these observations suggest that MOAP1 serves a distinct role in facilitating autophagy through interacting with LC3 to promote efficient phagophore closure during starvation.Abbreviations: CQ: Chloroquine; EBSS: Earle's Balanced Salt Solution; GABARAP: Gamma-Amino Butyric Acid Receptor Associated Protein; IF: Immunofluorescence; IP: Immunoprecipitation; LAMP1: Lysosomal-Associated Membrane Protein 1; LIR: LC3-Interacting Region; MAP1LC3/LC3: Microtubule Associated Protein 1 Light Chain 3; MEF: Mouse Embryonic Fibroblast; MOAP1: Modulator of Apoptosis 1; PE: Phosphatidylethanolamine; PtdIns3K: class III PtdIns3K complex I; PtdIns3P: Phosphatidylinositol-3-phosphate; STX17: Syntaxin 17; ULK1: unc-51 like autophagy activating kinase 1.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Roche
Protein A Agarose, >98% (HPLC and SDS-PAGE), suspension
Sigma-Aldrich
Anti-beta-Actin antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP124, affinity isolated antibody
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Bafilomycin A1 Ready Made Solution, 0.16 mM in DMSO, from Streptomyces griseus
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture