Merck
  • The Ca2+ -binding protein sorcin stimulates transcriptional activity of the unfolded protein response mediator ATF6.
The Ca2+ -binding protein sorcin stimulates transcriptional activity of the unfolded protein response mediator ATF6.
FEBS letters (2021-05-08)
Steven Z Parks, Tian Gao, Natalia Jimenez Awuapura, Joseph Ayathamattam, Pauline L Chabosseau, Dhananjaya V Kalvakolanu, Héctor H Valdivia, Guy A Rutter, Isabelle Leclerc
ABSTRACT
Sorcin is a calcium-binding protein involved in maintaining endoplasmic reticulum (ER) Ca2+ stores. We have previously shown that overexpressing sorcin under the rat insulin promoter was protective against high-fat diet-induced pancreatic beta-cell dysfunction in vivo. Activating transcription factor 6 (ATF6) is a key mediator of the unfolded protein response (UPR) that provides cellular protection during the progression of ER stress. Here, using nonexcitable HEK293 cells, we show that sorcin overexpression increased ATF6 signalling, whereas sorcin knock out caused a reduction in ATF6 transcriptional activity and increased ER stress. Altogether, our data suggest that sorcin downregulation during lipotoxic stress may prevent full ATF6 activation and a normal UPR during the progression of obesity and insulin resistance.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Green Fluorescent Protein (GFP), N-terminal antibody, Mouse monoclonal, clone GSN24, purified from hybridoma cell culture
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
Thapsigargin - CAS 67526-95-8 - Calbiochem, Thapsigargin, CAS 67526-95-8, is a cell-permeable, tumor-promoting sesquiterpene lactone that releases calcium by non-competitvley inhibiting endoplasmic reticular Ca2+-ATPase (IC50 = 4-13 nM).