Merck
  • GPR17 expressing NG2-Glia: Oligodendrocyte progenitors serving as a reserve pool after injury.
GPR17 expressing NG2-Glia: Oligodendrocyte progenitors serving as a reserve pool after injury.
Glia (2015-10-16)
Francesca Viganò, Sarah Schneider, Mauro Cimino, Elisabetta Bonfanti, Paolo Gelosa, Luigi Sironi, Maria P Abbracchio, Leda Dimou
ABSTRACT
In the adult brain NG2-glia continuously generate mature, myelinating oligodendrocytes. To which extent the differentiation process is common to all NG2-glia and whether distinct pools are recruited for repair under physiological and pathological conditions still needs clarification. Here, we aimed at investigating the differentiation potential of adult NG2-glia that specifically express the G-protein coupled receptor 17 (GPR17), a membrane receptor that regulates the differentiation of these cells at postnatal stages. To this aim, we generated the first BAC transgenic GPR17-iCreER(T2) mouse line for fate mapping studies. In these mice, under physiological conditions, GPR17(+) cells--in contrast to GPR17(-) NG2-glia--did not differentiate within 3 months, a peculiarity that was overcome after cerebral damage induced by acute injury or ischemia. After these insults, GPR17(+) NG2-glia rapidly reacted to the damage and underwent maturation, suggesting that they represent a 'reserve pool' of adult progenitors maintained for repair purposes.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Glial Fibrillary Acidic Protein (GFAP) antibody produced in mouse, clone G-A-5, ascites fluid