Merck
  • NRF1 association with AUTS2-Polycomb mediates specific gene activation in the brain.
NRF1 association with AUTS2-Polycomb mediates specific gene activation in the brain.
Molecular cell (2021-10-13)
Sanxiong Liu, Kimberly A Aldinger, Chi Vicky Cheng, Takae Kiyama, Mitali Dave, Hanna K McNamara, Wukui Zhao, James M Stafford, Nicolas Descostes, Pedro Lee, Stefano G Caraffi, Ivan Ivanovski, Edoardo Errichiello, Christiane Zweier, Orsetta Zuffardi, Michael Schneider, Antigone S Papavasiliou, M Scott Perry, Jennifer Humberson, Megan T Cho, Astrid Weber, Andrew Swale, Tudor C Badea, Chai-An Mao, Livia Garavelli, William B Dobyns, Danny Reinberg
ABSTRACT
The heterogeneous family of complexes comprising Polycomb repressive complex 1 (PRC1) is instrumental for establishing facultative heterochromatin that is repressive to transcription. However, two PRC1 species, ncPRC1.3 and ncPRC1.5, are known to comprise novel components, AUTS2, P300, and CK2, that convert this repressive function to that of transcription activation. Here, we report that individuals harboring mutations in the HX repeat domain of AUTS2 exhibit defects in AUTS2 and P300 interaction as well as a developmental disorder reflective of Rubinstein-Taybi syndrome, which is mainly associated with a heterozygous pathogenic variant in CREBBP/EP300. Moreover, the absence of AUTS2 or mutation in its HX repeat domain gives rise to misregulation of a subset of developmental genes and curtails motor neuron differentiation of mouse embryonic stem cells. The transcription factor nuclear respiratory factor 1 (NRF1) has a novel and integral role in this neurodevelopmental process, being required for ncPRC1.3 recruitment to chromatin.
Sigma-Aldrich
Tamoxifen, ≥99%
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sigma-Aldrich
Anti-DEDAF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution