Merck
  • Replating Protocol for Human Induced Pluripotent Stem Cell-Derived Cardiomyocytes.
Replating Protocol for Human Induced Pluripotent Stem Cell-Derived Cardiomyocytes.
Methods in molecular biology (Clifton, N.J.) (2021-12-01)
Arzuhan Koc, Esra Cagavi
ABSTRACT
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CM) create an unlimited cell source for basic and translational cardiac research. Obtaining hiPSC-CM culture as a single-cell, monolayer or three-dimensional clusters for downstream applications can be challenging. Thus, it is critical to develop replating strategies for hiPSC-CMs by evaluating different enzymatic or nonenzymatic reagents for dissociation and seeding on different coating materials. To reseed hiPSC-CMs with high viability and at structures desirable for the downstream applications, here we defined optimized protocols to dissociate hiPSC-CMs by using collagenase A&B, Collagenase II, TrypLE, and EDTA and reseeding on various matrix materials including fibronectin, laminin, imatrix, Matrigel, and Geltrex. By the replating methods described here, a single cell or cluster-containing hiPSC-CM cultures can be generated effectively.
Sigma-Aldrich
CHIR99021, ≥98% (HPLC)
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
ROCK Inhibitor (Y-27632), The ROCK Inhibitor (Y-27632) is available in a 5 mg format & has been optimized & validated for cell culture & Neuroscience applications.