Merck
  • Intrinsic antiviral immunity of barrier cells revealed by an iPSC-derived blood-brain barrier cellular model.
Intrinsic antiviral immunity of barrier cells revealed by an iPSC-derived blood-brain barrier cellular model.
Cell reports (2022-06-02)
Yichen Cheng, Angelica Medina, Zhenlan Yao, Mausumi Basu, Janhavi P Natekar, Jianshe Lang, Egan Sanchez, Mezindia B Nkembo, Chongchong Xu, Xuyu Qian, Phuong T T Nguyen, Zhexing Wen, Hongjun Song, Guo-Li Ming, Mukesh Kumar, Margo A Brinton, Melody M H Li, Hengli Tang
ABSTRACT
Physiological blood-tissue barriers play a critical role in separating the circulation from immune-privileged sites and denying access to blood-borne viruses. The mechanism of virus restriction by these barriers is poorly understood. We utilize induced pluripotent stem cell (iPSC)-derived human brain microvascular endothelial cells (iBMECs) to study virus-blood-brain barrier (BBB) interactions. These iPSC-derived cells faithfully recapitulate a striking difference in in vivo neuroinvasion by two alphavirus isolates and are selectively permissive to neurotropic flaviviruses. A model of cocultured iBMECs and astrocytes exhibits high transendothelial electrical resistance and blocks non-neurotropic flaviviruses from getting across the barrier. We find that iBMECs constitutively express an interferon-induced gene, IFITM1, which preferentially restricts the replication of non-neurotropic flaviviruses. Barrier cells from blood-testis and blood-retinal barriers also constitutively express IFITMs that contribute to the viral resistance. Our application of a renewable human iPSC-based model for studying virus-BBB interactions reveals that intrinsic immunity at the barriers contributes to virus exclusion.
Sigma-Aldrich
Anti-dsRNA Antibody, clone rJ2, culture supernatant, clone rJ2, from mouse
Sigma-Aldrich
Cyclosporin A, Ready Made Solution, 1 mg/mL in DMSO
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture, ≥95% (SDS-PAGE)
Sigma-Aldrich
Collagen Type I, rat tail, 100 mg, solution, BioReagent, suitable for cell culture