Merck
  • Age-induced prostaglandin E2 impairs mitochondrial fitness and increases mortality to influenza infection.
Age-induced prostaglandin E2 impairs mitochondrial fitness and increases mortality to influenza infection.
Nature communications (2022-11-10)
Judy Chen, Jane C Deng, Rachel L Zemans, Karim Bahmed, Beata Kosmider, Min Zhang, Marc Peters-Golden, Daniel R Goldstein
ABSTRACT
Aging impairs the immune responses to influenza A virus (IAV), resulting in increased mortality to IAV infections in older adults. However, the factors within the aged lung that compromise host defense to IAV remain unknown. Using a murine model and human samples, we identified prostaglandin E2 (PGE2), as such a factor. Senescent type II alveolar epithelial cells (AECs) are overproducers of PGE2 within the aged lung. PGE2 impairs the proliferation of alveolar macrophages (AMs), critical cells for defense against respiratory pathogens, via reduction of oxidative phosphorylation and mitophagy. Importantly, blockade of the PGE2 receptor EP2 in aged mice improves AM mitochondrial function, increases AM numbers and enhances survival to IAV infection. In conclusion, our study reveals a key mechanism that compromises host defense to IAV, and possibly other respiratory infections, with aging and suggests potential new therapeutic or preventative avenues to protect against viral respiratory disease in older adults.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Roche
Collagenase D, from Clostridium histolyticum
Roche
DNase I recombinant, grade I, from bovine pancreas, expressed in Pichia pastoris
Sigma-Aldrich
Dasatinib, ≥98% (HPLC)