Merck
  • Caveolin-1 dolines form a distinct and rapid caveolae-independent mechanoadaptation system.
Caveolin-1 dolines form a distinct and rapid caveolae-independent mechanoadaptation system.
Nature cell biology (2022-12-22)
Fidel-Nicolás Lolo, Nikhil Walani, Eric Seemann, Dobryna Zalvidea, Dácil María Pavón, Gheorghe Cojoc, Moreno Zamai, Christine Viaris de Lesegno, Fernando Martínez de Benito, Miguel Sánchez-Álvarez, Juan José Uriarte, Asier Echarri, Daniel Jiménez-Carretero, Joan-Carles Escolano, Susana A Sánchez, Valeria R Caiolfa, Daniel Navajas, Xavier Trepat, Jochen Guck, Christophe Lamaze, Pere Roca-Cusachs, Michael M Kessels, Britta Qualmann, Marino Arroyo, Miguel A Del Pozo
ABSTRACT
In response to different types and intensities of mechanical force, cells modulate their physical properties and adapt their plasma membrane (PM). Caveolae are PM nano-invaginations that contribute to mechanoadaptation, buffering tension changes. However, whether core caveolar proteins contribute to PM tension accommodation independently from the caveolar assembly is unknown. Here we provide experimental and computational evidence supporting that caveolin-1 confers deformability and mechanoprotection independently from caveolae, through modulation of PM curvature. Freeze-fracture electron microscopy reveals that caveolin-1 stabilizes non-caveolar invaginations-dolines-capable of responding to low-medium mechanical forces, impacting downstream mechanotransduction and conferring mechanoprotection to cells devoid of caveolae. Upon cavin-1/PTRF binding, doline size is restricted and membrane buffering is limited to relatively high forces, capable of flattening caveolae. Thus, caveolae and dolines constitute two distinct albeit complementary components of a buffering system that allows cells to adapt efficiently to a broad range of mechanical stimuli.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone MB1.2, clone MB1.2, Chemicon®, from rat
Sigma-Aldrich
Monoclonal Anti-Caveolin-1 antibody produced in mouse, clone CAV1, tissue culture supernatant