Merck
  • The ribose methylation enzyme FTSJ1 has a conserved role in neuron morphology and learning performance.
The ribose methylation enzyme FTSJ1 has a conserved role in neuron morphology and learning performance.
Life science alliance (2023-02-01)
Mira Brazane, Dilyana G Dimitrova, Julien Pigeon, Chiara Paolantoni, Tao Ye, Virginie Marchand, Bruno Da Silva, Elise Schaefer, Margarita T Angelova, Zornitza Stark, Martin Delatycki, Tracy Dudding-Byth, Jozef Gecz, Pierre-Yves Plaçais, Laure Teysset, Thomas Préat, Amélie Piton, Bassem A Hassan, Jean-Yves Roignant, Yuri Motorin, Clément Carré
ABSTRACT
FTSJ1 is a conserved human 2'-O-methyltransferase (Nm-MTase) that modifies several tRNAs at position 32 and the wobble position 34 in the anticodon loop. Its loss of function has been linked to X-linked intellectual disability (XLID), and more recently to cancers. However, the molecular mechanisms underlying these pathologies are currently unclear. Here, we report a novel FTSJ1 pathogenic variant from an X-linked intellectual disability patient. Using blood cells derived from this patient and other affected individuals carrying FTSJ1 mutations, we performed an unbiased and comprehensive RiboMethSeq analysis to map the ribose methylation on all human tRNAs and identify novel targets. In addition, we performed a transcriptome analysis in these cells and found that several genes previously associated with intellectual disability and cancers were deregulated. We also found changes in the miRNA population that suggest potential cross-regulation of some miRNAs with these key mRNA targets. Finally, we show that differentiation of FTSJ1-depleted human neural progenitor cells into neurons displays long and thin spine neurites compared with control cells. These defects are also observed in Drosophila and are associated with long-term memory deficits. Altogether, our study adds insight into FTSJ1 pathologies in humans and flies by the identification of novel FTSJ1 targets and the defect in neuron morphology.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Anti-Sox2 Antibody, Chemicon®, from rabbit