Merck
  • Follicle-Stimulating Hormone Glycosylation Variants Distinctly Modulate Pre-antral Follicle Growth and Survival.
Follicle-Stimulating Hormone Glycosylation Variants Distinctly Modulate Pre-antral Follicle Growth and Survival.
Endocrinology (2022-10-07)
Gillian P Johnson, Caitlan G A Onabanjo, Kate Hardy, Viktor Y Butnev, George R Bousfield, Kim C Jonas
ABSTRACT
Follicle-stimulating hormone (FSH) is a key endocrine regulator of ovarian function. FSH is secreted as 2 macroglycosylation variants: partially glycosylated FSH (FSH21/18) and fully glycosylated FSH (FSH24). FSH21/18 is more potent than FSH24 at binding to and activating the FSH receptor (R). The ratio of FSH21/18:FSH24 has been shown to change with age, with FSH21/18 predominant at reproductive prime, and FSH24 predominant during perimenopause/menopause. How these FSH glycosylation variants modulate ovarian follicle functions remains largely unknown. The aim of this study was to investigate the effect of FSH glycosylation variants of pre-antral follicle function. Pre-antral follicles were isolated from 3- to 5-week-old C57BL/6 mice and treated ±10 ng/mL FSH21/18, FSH24, a ratio of 80:20 FSH21/18:FSH24 (to mimic reproductive prime), 50:50 FSH21/18:FSH24 (perimenopause), or 20:80 FSH21/18:FSH24 (menopause) for up to 96 hours. FSH21/18 and 80:20 FSH21/18:FSH24 increased follicle growth, in comparison with control, contrasting with FSH24 and 20:80 FSH21/18:FSH24. Survival rates were decreased in follicles treated with FSH24 or 20:80 FSH21/18:FSH24, with follicles undergoing basement membrane rupture and oocyte extrusion, increased Caspase3 gene and protein expression, and decreased markers of cell proliferation in FSH24 or 20:80 FSH21/18:FSH24-treated follicles. Moreover, this correlated with differential regulation of key genes modulating follicular functions. Pharmacological inhibitors of key FSH signal pathways suggests FSH21/18 and FSH24 initiate different FSHR signal pathway activation, which may determine their differential effects on follicle growth and survival. These data suggest that the nature of FSH glycosylation modulates the follicular cellular environment to regulate follicle growth and survival and may underpin the increasing ovarian resistance to FSH observed during aging.
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)