Merck
  • Human-specific regulation of neural maturation identified by cross-primate transcriptomics.
Human-specific regulation of neural maturation identified by cross-primate transcriptomics.
Current biology : CB (2022-10-14)
Sara B Linker, Iñigo Narvaiza, Jonathan Y Hsu, Meiyan Wang, Fan Qiu, Ana P D Mendes, Ruth Oefner, Kalyani Kottilil, Amandeep Sharma, Lynne Randolph-Moore, Eunice Mejia, Renata Santos, Maria C Marchetto, Fred H Gage
ABSTRACT
Unique aspects of human behavior are often attributed to differences in the relative size and organization of the human brain: these structural aspects originate during early development. Recent studies indicate that human neurodevelopment is considerably slower than that in other nonhuman primates, a finding that is termed neoteny. One aspect of neoteny is the slow onset of action potentials. However, which molecular mechanisms play a role in this process remain unclear. To examine the evolutionary constraints on the rate of neuronal maturation, we have generated transcriptional data tracking five time points, from the neural progenitor state to 8-week-old neurons, in primates spanning the catarrhine lineage, including Macaca mulatta, Gorilla gorilla, Pan paniscus, Pan troglodytes, and Homo sapiens. Despite finding an overall similarity of many transcriptional signatures, species-specific and clade-specific distinctions were observed. Among the genes that exhibited human-specific regulation, we identified a key pioneer transcription factor, GATA3, that was uniquely upregulated in humans during the neuronal maturation process. We further examined the regulatory nature of GATA3 in human cells and observed that downregulation quickened the speed of developing spontaneous action potentials, thereby modulating the human neotenic phenotype. These results provide evidence for the divergence of gene regulation as a key molecular mechanism underlying human neoteny.
Sigma-Aldrich
Anti-Nestin, Human Antibody, serum, from rabbit