Merck
  • Plastin 3 rescues cell surface translocation and activation of TrkB in spinal muscular atrophy.
Plastin 3 rescues cell surface translocation and activation of TrkB in spinal muscular atrophy.
The Journal of cell biology (2023-01-07)
Luisa Hennlein, Hanaa Ghanawi, Florian Gerstner, Eduardo Palominos García, Ezgi Yildirim, Lena Saal-Bauernschubert, Mehri Moradi, Chunchu Deng, Teresa Klein, Silke Appenzeller, Markus Sauer, Michael Briese, Christian Simon, Michael Sendtner, Sibylle Jablonka
ABSTRACT
Plastin 3 (PLS3) is an F-actin-bundling protein that has gained attention as a modifier of spinal muscular atrophy (SMA) pathology. SMA is a lethal pediatric neuromuscular disease caused by loss of or mutations in the Survival Motor Neuron 1 (SMN1) gene. Pathophysiological hallmarks are cellular maturation defects of motoneurons prior to degeneration. Despite the observed beneficial modifying effect of PLS3, the mechanism of how it supports F-actin-mediated cellular processes in motoneurons is not yet well understood. Our data reveal disturbed F-actin-dependent translocation of the Tropomyosin receptor kinase B (TrkB) to the cell surface of Smn-deficient motor axon terminals, resulting in reduced TrkB activation by its ligand brain-derived neurotrophic factor (BDNF). Improved actin dynamics by overexpression of hPLS3 restores membrane recruitment and activation of TrkB and enhances spontaneous calcium transients by increasing Cav2.1/2 "cluster-like" formations in SMA axon terminals. Thus, our study provides a novel role for PLS3 in supporting correct alignment of transmembrane proteins, a key mechanism for (moto)-neuronal development.
Sigma-Aldrich
Cytochalasin D, Ready Made Solution, from Zygosporium mansonii, 5 mg/mL in DMSO
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Anti-Neurofilament H Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-ARP3 antibody, Mouse monoclonal, clone FMS338, purified from hybridoma cell culture
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat