Merck
  • Synthesis of Indole-Fused Dihydrothiopyrano Scaffolds via (3 + 3)-Annulations of Donor-Acceptor Cyclopropanes with Indoline-2-Thiones.
Synthesis of Indole-Fused Dihydrothiopyrano Scaffolds via (3 + 3)-Annulations of Donor-Acceptor Cyclopropanes with Indoline-2-Thiones.
The Journal of organic chemistry (2022-12-17)
Braj Gopal, Prasoon Raj Singh, Madan Kumar, Avijit Goswami
ABSTRACT
A new methodology for the synthesis of N-haloindole-fused dihydrothiopyrano derivatives via (3 + 3)-annulation of donor-acceptor cyclopropanes (DACs) with indoline-2-thiones in the presence of Sc(OTf)3 as a Lewis acid catalyst has been developed. This protocol provides a variety of indole-fused dihydrothiopyrano molecules in good to excellent yields, which architecturally resemble other indole-fused tricyclic molecules having potential medicinal value. In addition, we have described a detailed reaction mechanism and transformation of the furnished product into N-fused thiazino indole molecule.
Sigma-Aldrich
Indoline-2-thione, ≥-95%