Merck
  • Development and validation of an LC-MS assay for finasteride and its application to prostate cancer prevention trial sample analysis.
Development and validation of an LC-MS assay for finasteride and its application to prostate cancer prevention trial sample analysis.
Journal of chromatographic science (2008-04-12)
Xiaohong Chen, Erin R Gardner, Douglas K Price, William D Figg
ABSTRACT
An analytical method is developed and validated for the quantitative determination of finasteride, a potent 5 alpha-reductase inhibitor, in human plasma. Calibration curves are linear in the concentration range of 1 to 100 ng/mL. Sample pretreatment involves a liquid-liquid extraction with ethyl acetate using 0.2 mL aliquots of plasma. Finasteride and the internal standard (beclomethasone) are separated on a Waters Symmetry Shield RP18 column (50 x 2.1 mm, 3.5 microm) and eluted using a gradient mobile phase composed of acetonitrile and 10mM ammonium acetate with 0.1% formic acid. The column eluant is monitored by mass spectrometry with electrospray ionization. A complete validation of the method is performed. For quality control samples at three different concentrations that were analyzed in quintuplicate, on six separate occasions, the accuracy and precision range from 95.2% to 101% and 3.4% to 7.3%, respectively. The developed method is subsequently applied to measure the steady state finasteride concentration of patients who participated in the Prostate Cancer Prevention Trial.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%