Merck
  • Nicotinamide Antagonizes Lipopolysaccharide-Induced Hypoxic Cell Signals in Human Macrophages.
Nicotinamide Antagonizes Lipopolysaccharide-Induced Hypoxic Cell Signals in Human Macrophages.
Journal of immunology (Baltimore, Md. : 1950) (2023-06-14)
Colleen S Curran, Edward J Dougherty, Xizhong Cui, Yan Li, Mark Jeakle, Tom Gamble, Cumhur Y Demirkale, Parizad Torabi-Parizi
ABSTRACT
Mechanisms to control the immune response are important to pathogen evasion and host defense. Gram-negative bacteria are common pathogens that can activate host immune responses through their outer membrane component, LPS. Macrophage activation by LPS induces cell signals that promote hypoxic metabolism, phagocytosis, Ag presentation, and inflammation. Nicotinamide (NAM) is a vitamin B3 derivative and precursor in the formation of NAD, which is a required cofactor in cellular function. In this study, treatment of human monocyte-derived macrophages with NAM promoted posttranslational modifications that antagonized LPS-induced cell signals. Specifically, NAM inhibited AKT and FOXO1 phosphorylation, decreased p65/RelA acetylation, and promoted p65/RelA and hypoxia-inducible transcription factor-1α (HIF-1α) ubiquitination. NAM also increased prolyl hydroxylase domain 2 (PHD2) production, inhibited HIF-1α transcription, and promoted the formation of the proteasome, resulting in reduced HIF-1α stabilization, decreased glycolysis and phagocytosis, and reductions in NOX2 activity and the production of lactate dehydrogenase A. These NAM responses were associated with increased intracellular NAD levels formed through the salvage pathway. NAM and its metabolites may therefore decrease the inflammatory response of macrophages and protect the host against excessive inflammation but potentially increase injury through reduced pathogen clearance. Continued study of NAM cell signals in vitro and in vivo may provide insight into infection-associated host pathologies and interventions.
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
Mouse IgG−Agarose, (Suspension in 0.5 M NaCl containing preservative.)
Sigma-Aldrich
Anti-PSMF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%