Merck
  • STAT3 palmitoylation initiates a positive feedback loop that promotes the malignancy of hepatocellular carcinoma cells in mice.
STAT3 palmitoylation initiates a positive feedback loop that promotes the malignancy of hepatocellular carcinoma cells in mice.
Science signaling (2023-12-06)
Yi Jiang, Yuejie Xu, Chengliang Zhu, Guifang Xu, Lei Xu, Zijian Rao, Lixing Zhou, Ping Jiang, Sara Malik, Jingyuan Fang, Hening Lin, Mingming Zhang
ABSTRACT
Constitutive activation of the transcription factor STAT3 (signal transducer and activator of transcription 3) contributes to the malignancy of many cancers such as hepatocellular carcinoma (HCC) and is associated with poor prognosis. STAT3 activity is increased by the reversible palmitoylation of Cys108 by the palmitoyltransferase DHHC7 (encoded by ZDHHC7). Here, we investigated the consequences of S-palmitoylation of STAT3 in HCC. Increased ZDHHC7 abundance in HCC cases was associated with poor prognosis, as revealed by bioinformatics analysis of patient data. In HepG2 cells in vitro, DHHC7-mediated palmitoylation enhanced the expression of STAT3 target genes, including HIF1A, which encodes the hypoxia-inducible transcription factor HIF1α. Inhibiting DHHC7 decreased the S-palmitoylation of STAT3 and decreased HIF1α abundance. Furthermore, stabilization of HIF1α by cyclin-dependent kinase 5 (CDK5) enabled it to promote the expression of ZDHHC7, which generated a positive feedback loop between DHHC7, STAT3, and HIF1α. Perturbing this loop reduced the growth of HCC cells in vivo. Moreover, DHHC7, STAT3, and HIF1α were all abundant in human HCC tissues. Our study identifies a pathway connecting these proteins that is initiated by S-palmitoylation, which may be broadly applicable to understanding the role of this modification in cancer.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Millipore
EZview Red Anti-HA Affinity Gel
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%