Merck
  • MYSM1 acts as a novel co-activator of ERα to confer antiestrogen resistance in breast cancer.
MYSM1 acts as a novel co-activator of ERα to confer antiestrogen resistance in breast cancer.
EMBO molecular medicine (2024-01-05)
Ruina Luan, Mingcong He, Hao Li, Yu Bai, Anqi Wang, Ge Sun, Baosheng Zhou, Manlin Wang, Chunyu Wang, Shengli Wang, Kai Zeng, Jianwei Feng, Lin Lin, Yuntao Wei, Shigeaki Kato, Qiang Zhang, Yue Zhao
ABSTRACT
Endocrine resistance is a crucial challenge in estrogen receptor alpha (ERα)-positive breast cancer (BCa). Aberrant alteration in modulation of E2/ERα signaling pathway has emerged as the putative contributor for endocrine resistance in BCa. Herein, we demonstrate that MYSM1 as a deubiquitinase participates in modulating ERα action via histone and non-histone deubiquitination. MYSM1 is involved in maintenance of ERα stability via ERα deubiquitination. MYSM1 regulates relevant histone modifications on cis regulatory elements of ERα-regulated genes, facilitating chromatin decondensation. MYSM1 is highly expressed in clinical BCa samples. MYSM1 depletion attenuates BCa-derived cell growth in xenograft models and increases the sensitivity of antiestrogen agents in BCa cells. A virtual screen shows that the small molecule Imatinib could potentially interact with catalytic MPN domain of MYSM1 to inhibit BCa cell growth via MYSM1-ERα axis. These findings clarify the molecular mechanism of MYSM1 as an epigenetic modifier in regulation of ERα action and provide a potential therapeutic target for endocrine resistance in BCa.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit